missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylenedicarboxylate, 98%

Product Code. 11450293
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450293 25 g 25g
11460293 100 g 100g
11470293 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450293 Supplier Thermo Scientific Alfa Aesar Supplier No. A11437.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 762-42-5 | C6H6O4 | 142.11 g/mol

It acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.

Solubility
It is insoluble in water but soluble in ethanol, ethyl ether, CCl4, miscible with organic solvents.

Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents, strong acids, strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-42-5
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00008456
InChI Key VHILMKFSCRWWIJ-UHFFFAOYSA-N
Synonym dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene
PubChem CID 12980
IUPAC Name dimethyl but-2-ynedioate
SMILES COC(=O)C#CC(=O)OC

Specifications

Melting Point -18°C
Density 1.156
Boiling Point 95°C to 98°C (19 mmHg)
Flash Point 86°C (186°F)
Linear Formula CH3O2CC≡CCO2CH3
Refractive Index 1.448
Quantity 25 g
UN Number UN1760
Beilstein 607063
Sensitivity Light sensitive
Solubility Information It is insoluble in water but soluble in ethanol,ethyl ether,CCl4,miscible with organic solvents.
Formula Weight 142.11
Percent Purity 98%
Chemical Name or Material Dimethyl acetylenedicarboxylate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl acetylenedicarboxylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Butynedioic acid,dimethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.