missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylenedicarboxylate, 95%

Product Code. 15412167
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412167 25 g 25g
15422167 100 g 100g
15432167 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412167 Supplier Thermo Scientific Alfa Aesar Supplier No. 045131.14
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 762-42-5 | C6H6O4 | 142.11 g/mol

Dimethyl acetylenedicarboxylate as a versatile dienophile used in Diels-Alder reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-42-5
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00008456
InChI Key VHILMKFSCRWWIJ-UHFFFAOYSA-N
Synonym dimethyl acetylenedicarboxylate, 2-butynedioic acid, dimethyl ester, 1,4-dimethyl but-2-ynedioate, dimethyl butynedioate, dimethyl 2-butynedioate, acetylenedicarboxylic acid dimethyl ester, dmad, methyl acetylenedicarboxylate, bis methoxycarbonyl acetylene, di carbomethoxy acetylene
PubChem CID 12980
IUPAC Name dimethyl but-2-ynedioate
SMILES COC(=O)C#CC(=O)OC

Specifications

Melting Point -18°C
Color Colorless to Yellow
Density 1.156 g/mL
Boiling Point 95°C to 98°C (19 mmHg)
Flash Point 86°C (186°F)
Linear Formula CH3O2CC≡CCO2CH3
Refractive Index 1.447
Quantity 25 g
UN Number UN1760
Beilstein 607063
Sensitivity Light sensitive
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 142.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dimethyl acetylenedicarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.