missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetone-1,3-dicarboxylate, 97%

Product Code. 11427177
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427177 500 g 500g
11407177 25 g 25g
11417177 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11427177 Supplier Thermo Scientific Alfa Aesar Supplier No. A14969.36
 Restricted Item
In Stock
Add to basket
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto es un intermedio aislado transportado de acuerdo con el Reglamento (CE) 1907/2006 (REACH). Todos los clientes deben completar una declaración antes de la entrega.

CAS: 1830-54-2 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00008462
InChI Key RNJOKCPFLQMDEC-UHFFFAOYSA-N
Synonym dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
PubChem CID 74591
SMILES COC(=O)CC(=O)CC(=O)OC

Especificaciones

Melting Point 16°C to 17°C
Density 1.185
Boiling Point 237°C to 241°C
Flash Point >110°C (230°F)
Odor Ester-like
Refractive Index 1.444
Quantity 500 g
Beilstein 1366042
IUPAC Name dimethyl 3-oxopentanedioate
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Dimethyl acetone-1,3-dicarboxylate
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.