missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetone-1,3-dicarboxylate, 97%

Product Code. 11417177
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11417177

Brand: Thermo Scientific Alfa Aesar A14969.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 1830-54-2 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00008462
InChI Key RNJOKCPFLQMDEC-UHFFFAOYSA-N
Synonym dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
PubChem CID 74591
SMILES COC(=O)CC(=O)CC(=O)OC

Specifications

Melting Point 16°C to 17°C
Density 1.185
Boiling Point 237°C to 241°C
Flash Point >110°C (230°F)
Odor Ester-like
Refractive Index 1.444
Quantity 100 g
Beilstein 1366042
IUPAC Name dimethyl 3-oxopentanedioate
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Dimethyl acetone-1,3-dicarboxylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.