missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-sulfoisophthalate sodium salt, 98%, Thermo Scientific™

Product Code. 11450508
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450508 25 g 25g
11460508 100 g 100g
11470508 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450508 Supplier Thermo Scientific Alfa Aesar Supplier No. B24646.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3965-55-7
Molecular Formula C10H9NaO7S
Molecular Weight (g/mol) 296.225
MDL Number MFCD00007493
InChI Key LLHSEQCZSNZLRI-UHFFFAOYSA-M
Synonym sodium 3,5-bis methoxycarbonyl benzenesulfonate, sodium dimethyl 5-sulphonatoisophthalate, dimethyl 5-sulfoisophthalate sodium salt, unii-7l3g5x9vdu, sodium 1,3-dimethyl 5-sulfoisophthalate, 7l3g5x9vdu, 5-sulfoisophthalic acid, dimethyl ester, sodium salt, sodium dimethyl 5-sulfoisophthalate, 1,3-benzenedicarboxylic acid, 5-sulfo-, 1,3-dimethyl ester, sodium salt, 3,5-bis-methylkarboxy-benzensulfonan sodny czech
PubChem CID 23675796
IUPAC Name sodium;3,5-bis(methoxycarbonyl)benzenesulfonate
SMILES COC(=O)C1=CC(=CC(=C1)S(=O)(=O)[O-])C(=O)OC.[Na+]

Specifications

Melting Point >300°C
Quantity 25 g
Formula Weight 296.23
Percent Purity 98%
Chemical Name or Material Dimethyl 5-sulfoisophthalate sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.