missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5,5'-(Pyrazine-2,5-diyl)bis(2-hydroxybenzoate), TRC

Product Code. 30388718
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388718 5 mg 5mg
30380335 2.5 mg 2.5mg
30372512 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388718 Supplier Toronto Research Chemicals Supplier No. TRCD5220305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl 5,5'-(Pyrazine-2,5-diyl)bis(2-hydroxybenzoate)
Analyte or Component Names Dimethyl 5,5'-(Pyrazine-2,5-diyl)bis(2-hydroxybenzoate)
Shipping Condition Room Temperature
Molecular Formula C20 H16 N2 O6
InChI Formula InChI=1S/C20H16N2O6/c1-27-19(25)13-7-11(3-5-17(13)23)15-9-22-16(10-21-15)12-4-6-18(24)14(8-12)20(26)28-2/h3-10,23-24H,1-2H3
Solution Type Neat
SMILES COC(=O)c1cc(ccc1O)c2cnc(cn2)c3ccc(O)c(c3)C(=O)OC
IUPAC Name methyl 2-hydroxy-5-[5-(4-hydroxy-3-methoxycarbonylphenyl)pyrazin-2-yl]benzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 380.351
Quantity 5 mg
Formula Weight 380.101
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.