missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%

Product Code. 11460491
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460491 5 g 5g
11450491 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460491 Supplier Thermo Scientific Alfa Aesar Supplier No. B20028.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121071-71-4 | C8H10ClNO4S | 251.681 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 121071-71-4
Molecular Formula C8H10ClNO4S
Molecular Weight (g/mol) 251.681
MDL Number MFCD00205189
InChI Key RETIFGMAGNDLAN-UHFFFAOYSA-N
PubChem CID 2808333
IUPAC Name dimethyl 4-aminothiophene-2,3-dicarboxylate;hydrochloride
SMILES COC(=O)C1=C(SC=C1N)C(=O)OC.Cl

Spezifikation

Melting Point 166°C to 169°C
Quantity 5 g
Sensitivity Hygroscopic
Synonym dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 2,3-dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, acmc-209v6b, dimethyl 4-aminothiophene-2,3-dicarboxylate hcl, dimethyl 4-aminothiophene-2,3-dicarboxylate hydroc, 3-amino-4,5-bismethoxycarbonylthiophene hydrochloride, dimethyl4-aminothiophene-2,3-dicarboxylate hydrochloride, 4-aminothiophene-2,3-dicarboxylic acid dimethyl ester hydrochloride, methyl 4-amino-2-methoxycarbonyl thiophene-3-carboxylate, chloride
Formula Weight 251.69
Percent Purity 97%
Chemical Name or Material Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.