missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%

Product Code. 11450491
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11450491

Brand: Thermo Scientific Alfa Aesar B20028.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121071-71-4 | C8H10ClNO4S | 251.681 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121071-71-4
Molecular Formula C8H10ClNO4S
Molecular Weight (g/mol) 251.681
MDL Number MFCD00205189
InChI Key RETIFGMAGNDLAN-UHFFFAOYSA-N
PubChem CID 2808333
IUPAC Name dimethyl 4-aminothiophene-2,3-dicarboxylate;hydrochloride
SMILES COC(=O)C1=C(SC=C1N)C(=O)OC.Cl

Specifications

Melting Point 166°C to 169°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 2,3-dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, acmc-209v6b, dimethyl 4-aminothiophene-2,3-dicarboxylate hcl, dimethyl 4-aminothiophene-2,3-dicarboxylate hydroc, 3-amino-4,5-bismethoxycarbonylthiophene hydrochloride, dimethyl4-aminothiophene-2,3-dicarboxylate hydrochloride, 4-aminothiophene-2,3-dicarboxylic acid dimethyl ester hydrochloride, methyl 4-amino-2-methoxycarbonyl thiophene-3-carboxylate, chloride
Formula Weight 251.69
Percent Purity 97%
Chemical Name or Material Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.