missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 2-Isobutylmalonate, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Dimethyl 2-Isobutylmalonate |
| CAS | 39520-24-6 |
| Molecular Formula | C9H16O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H16O4/c1-6(2)5-7(8(10)12-3)9(11)13-4/h6-7H,5H2,1-4H3 |
| SMILES | COC(=O)C(CC(C)C)C(=O)OC |
| IUPAC Name | dimethyl 2-(2-methylpropyl)propanedioate |
| Molecular Weight (g/mol) | 188.22 |
| Formula Weight | 188.1049 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction