missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,6-Pyridinedicarboxylate, TRC

Product Code. 30388288
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30388288 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30388288

Brand: Toronto Research Chemicals TRCD47906025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl 2,6-Pyridinedicarboxylate
CAS 5453-67-8
Molecular Formula C9H9NO4
Quantity 25 g
Synonym 2,6-Pyridinedicarboxylic Acid 2,6-Dimethyl Ester, 2,6-Bismethoxycarbonylpyridine, 2,6-Dicarbomethoxypyridine, Dipicolinic Acid Dimethyl Ester, NSC 18855
InChI Formula InChI=1S/C9H9NO4/c1-13-8(11)6-4-3-5-7(10-6)9(12)14-2/h3-5H,1-2H3
SMILES COC(=O)c1cccc(n1)C(=O)OC
IUPAC Name dimethyl pyridine-2,6-dicarboxylate
Molecular Weight (g/mol) 195.17
Formula Weight 195.0532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.