missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,6-pyridinedicarboxylate, 99%

Product Code. 10734385 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10734385 50 g Glass bottle 50g
10071414 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10734385 Supplier Thermo Scientific Acros Supplier No. 427940500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 5453-67-8 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5453-67-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00134493
InChI Key SNQQJEJPJMXYTR-UHFFFAOYSA-N
Synonym dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester
PubChem CID 79549
IUPAC Name dimethyl pyridine-2,6-dicarboxylate
SMILES COC(=O)C1=NC(=CC=C1)C(=O)OC

Specifications

Melting Point 123°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 195.17
Percent Purity 99%
Chemical Name or Material Dimethyl 2, 6-pyridinedicarboxylate
compliance-icons compliance-icons
Product Identifier
  • Dimethyl 2,6-pyridinedicarboxylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.