missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dimethyl 2,3-O-isopropylidene-L-tartrate, TRC

Product Code. 30382655
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382655 25 g 25g
30381788 10 g 10g
30386555 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382655 Supplier Toronto Research Chemicals Supplier No. TRCD47424025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37031-29-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
Synonym (4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid 4,5-Dimethyl Ester, (4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid Dimethyl Ester, (4R-trans)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid Dimethyl Ester, (-)-Dimethyl-2,3-O-isopropylidene-L-tartrate, Dimethyl (4R,5R)-2,2,Dimethyl-1,3-dioxolane-4,5-dicarboxylate, Dimethyl L-tartarte Acetonide
SMILES COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H14O6/c1-9(2)14-5(7(10)12-3)6(15-9)8(11)13-4/h5-6H,1-4H3/t5-,6-/m1/s1
Formula Weight 218.079
Chemical Name or Material (-)-Dimethyl 2,3-O-isopropylidene-L-tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.