missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%

Product Code. 10677611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10677611 10 mL Glass bottle 10mL
10185833 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10677611 Supplier Thermo Scientific Acros Supplier No. 258440100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37031-30-4 | C9H14O6 | 218.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37031-30-4
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
MDL Number MFCD00040504
InChI Key ROZOUYVVWUTPNG-WDSKDSINSA-N
Synonym 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, +-dimethyl 2,3-o-isopropylidene-d-tartrate, dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4s,5s-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4,5-dimethyl 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6765, dimethyl--2,3-o-isopropylidene-l-tartrate, dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate #, +-dimethyl-2,3-o-isopropylidine-d-tartrate, dimethyl +-2,3-o-isopropylidene-d-tartrate
PubChem CID 688156
IUPAC Name dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
SMILES CC1(OC(C(O1)C(=O)OC)C(=O)OC)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 139°C (21.0 hPa)
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4386 to 1.4406
Quantity 10 mL
Specific Gravity 1.15
Specific Rotation Condition + 54.00 (20.00°C c=neat)
Specific Rotation + 54
Formula Weight 218.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (+)-Dimethyl 2, 3-O-isopropylidene-D-tartrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.