missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%

Product Code. 17162139 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17162139

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 90965-06-3 | C5H9N2O4P | 192.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90965-06-3
Molecular Formula C5H9N2O4P
Molecular Weight (g/mol) 192.11
InChI Key SQHSJJGGWYIFCD-UHFFFAOYSA-N
IUPAC Name 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate
SMILES COP(=O)(OC)C([N+]#N)=C(C)[O-]

Specifications

Assay Percent Range 95%
Quantity 5 g
Formula Weight 192.11
Chemical Name or Material Dimethyl (1-diazo-2-oxopropyl)phosphonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.