missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 98+%

Product Code. 10379480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10379480 500 g Glass bottle 500g
10580204 50 g Glass bottle 50g
10553411 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10379480 Supplier Thermo Scientific Acros Supplier No. 271815000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6289-46-9 | C10H12O6 | 228.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6289-46-9
Molecular Formula C10H12O6
Molecular Weight (g/mol) 228.20
MDL Number MFCD00001607
InChI Key MHKKFFHWMKEBDW-UHFFFAOYNA-N
Synonym dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
PubChem CID 94866
IUPAC Name dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
SMILES COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

Specifications

Melting Point 154°C to 157°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 500 g
Beilstein 10, IV, 3514
Formula Weight 228.2
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Dimethyl 1, 4-cyclohexanedione-2, 5-dicarboxylate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.