missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethylvinylsilane, 97%

Product Code. p-7046349
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11338915

Brand: Thermo Scientific Alfa Aesar L14035.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16753-62-1 | C5H12O2Si | 132.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16753-62-1
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00129170
InChI Key ZLNAFSPCNATQPQ-UHFFFAOYSA-N
Synonym vinylmethyldimethoxysilane, dimethoxymethylvinylsilane, dimethoxy methyl vinyl silane, silane, ethenyldimethoxymethyl, ethenyldimethoxymethylsilane, methyldimethoxyvinylsilane, vinyl methyldimethoxysilane, dimethoxy methyl vinylsilane #, methylvinyldimethoxysilane
PubChem CID 85589
IUPAC Name ethenyl-dimethoxy-methylsilane
SMILES CO[Si](C)(C=C)OC

Specifications

Boiling Point 102°C to 104°C
Quantity 5 g
UN Number UN1993
Beilstein 3043765
Flash Point 5°C (41°F)
Formula Weight 132.23
Percent Purity 97%
Refractive Index 1.395
Sensitivity Moisture sensitive
Density 0.889
Chemical Name or Material Dimethoxymethylvinylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.