missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, TRC

Product Code. 30377927
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377927 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377927 Supplier Toronto Research Chemicals Supplier No. TRCD460455250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethoxymethane
CAS 109-87-5
Molecular Formula C3 H8 O2
Quantity 250 mL
Synonym 1,1-Dimethoxymethane, 2,4-Dioxapentane, Anesthenyl, Bis(methoxy)methane, Dimethyl formal, Formal, Formaldehyde dimethyl acetal, Formaldehyde methyl ketal, Methoxymethyl methyl ether, Methylal, Methylene dimethyl ether, Methylene glycol dimethyl ether, ODS
InChI Formula InChI=1S/C3H8O2/c1-4-3-5-2/h3H2,1-2H3
SMILES COCOC
IUPAC Name dimethoxymethane
Molecular Weight (g/mol) 76.09
Formula Weight 76.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.