missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, TRC

Product Code. 30371919
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371919 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371919 Supplier Toronto Research Chemicals Supplier No. TRCD4604551L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethoxymethane
CAS 109-87-5
Molecular Formula C3 H8 O2
Quantity 1 L
Synonym 1,1-Dimethoxymethane, 2,4-Dioxapentane, Anesthenyl, Bis(methoxy)methane, Dimethyl formal, Formal, Formaldehyde dimethyl acetal, Formaldehyde methyl ketal, Methoxymethyl methyl ether, Methylal, Methylene dimethyl ether, Methylene glycol dimethyl ether, ODS
InChI Formula InChI=1S/C3H8O2/c1-4-3-5-2/h3H2,1-2H3
SMILES COCOC
IUPAC Name dimethoxymethane
Molecular Weight (g/mol) 76.09
Formula Weight 76.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.