missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxymethane, 99.5+%, Thermo Scientific Chemicals
44.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -105.0°C |
Color | Colorless |
Density | 0.8600g/mL |
Boiling Point | 41.0°C to 42.0°C |
Flash Point | −18°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH2(OCH3)2 |
Refractive Index | 1.3525 to 1.3545 |
Show More |
Safety and Handling
Product Identifier
- Dimethoxymethane
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'