missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, 98%

Product Code. 11499932
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11499932

Brand: Thermo Scientific Alfa Aesar A12055.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-87-5 | C3H8O2 | 76.095 g/mol

Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:

Solubility
Soluble in water 20°C 330g/L.

Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-87-5
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00008495
InChI Key NKDDWNXOKDWJAK-UHFFFAOYSA-N
Synonym methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
PubChem CID 8020
ChEBI CHEBI:48341
IUPAC Name dimethoxymethane
SMILES COCOC

Specifications

Melting Point -105°C
Density 0.863
Boiling Point 41°C to 42°C
Flash Point −17°C (1°F)
Linear Formula CH3OCH2OCH3
Refractive Index 1.3545
Quantity 2500 mL
UN Number UN1234
Beilstein 1697025
Sensitivity Moisture sensitive
Merck Index 14,6012
Solubility Information Soluble in water 20°C 330g/L.
Formula Weight 76.1
Percent Purity 98%
Chemical Name or Material Dimethoxymethane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.