missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxymethane, 98%
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
Description
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
Solubility
Soluble in water 20°C 330g/L.
Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:
Solubility
Soluble in water 20°C 330g/L.
Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
Specifications
Specifications
| Melting Point | -105°C |
| Density | 0.863 |
| Boiling Point | 41°C to 42°C |
| Flash Point | −17°C (1°F) |
| Linear Formula | CH3OCH2OCH3 |
| Refractive Index | 1.3545 |
| Quantity | 500 mL |
| UN Number | UN1234 |
| Beilstein | 1697025 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
Product Identifier
- Dimethoxymethane
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : Dimethoxymethane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction