missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydiphenylsilane, 97%

Product Code. 11386756
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11386756

Brand: Thermo Scientific Alfa Aesar L04031.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6843-66-9 | C14H16O2Si | 244.365 g/mol

Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6843-66-9
Molecular Formula C14H16O2Si
Molecular Weight (g/mol) 244.365
MDL Number MFCD00025690
InChI Key AHUXYBVKTIBBJW-UHFFFAOYSA-N
Synonym diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s
PubChem CID 81284
IUPAC Name dimethoxy(diphenyl)silane
SMILES CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC

Specifications

Boiling Point 160°C to 162°C (15mmHg)
Quantity 50 g
Beilstein 2940458
Flash Point 121°C (249°F)
Formula Weight 244.37
Percent Purity 97%
Refractive Index 1.542
Sensitivity Moisture sensitive
Density 1.079
Chemical Name or Material Dimethoxydiphenylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.