missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydimethylsilane, 97%

Product Code. 11489882
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489882 25 g 25g
11499882 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489882 Supplier Thermo Scientific Alfa Aesar Supplier No. A12025.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1112-39-6 | C4H12O2Si | 120.22 g/mol

It is employed as silylating agent for diols. Used as a monomer (‘building block’) in the production of silicone polymers or silicone resins, as an intermediate (starting material) in the production of other organosilicon substances. Also employed in non-metal surface treatment to enhance the adhesion properties of surfaces. Used in sealants, in silicone polymer preparations (for the production of elastomers), in mould-making, in semiconductor manufacture, as a laboratory chemical in research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as silylating agent for diols. Used as a monomer (‘building block’) in the production of silicone polymers or silicone resins, as an intermediate (starting material) in the production of other organosilicon substances. Also employed in non-metal surface treatment to enhance the adhesion properties of surfaces. Used in sealants, in silicone polymer preparations (for the production of elastomers), in mould-making, in semiconductor manufacture, as a laboratory chemical in research and development activities.

Solubility
Contact with water releases a flammable alcohol.

Notes
Moisture Sensitive. Store away from oxidizing agents and water/moisture. Protect against electrostatic charges. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-39-6
Molecular Formula C4H12O2Si
Molecular Weight (g/mol) 120.22
MDL Number MFCD00025691
InChI Key JJQZDUKDJDQPMQ-UHFFFAOYSA-N
Synonym dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
PubChem CID 66187
IUPAC Name dimethoxydimethylsilane
SMILES CO[Si](C)(C)OC

Specifications

Boiling Point 81°C to 82°C
Melting Point -80°C
Quantity 25 g
UN Number UN1993
Beilstein 1733694
Flash Point −8°C (17°F)
Solubility Information Contact with water releases a flammable alcohol.
Formula Weight 120.22
Percent Purity 97%
Refractive Index 1.371
Sensitivity Moisture sensitive
Density 0.867
Chemical Name or Material Dimethoxydimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.