missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxane, TRC

Product Code. 30373167
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373167 500 mg
30385472 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373167 Supplier Toronto Research Chemicals Supplier No. TRCD460495500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 828-00-2
Molecular Formula C8 H14 O4
Molecular Weight (g/mol) 174.19
Synonym 1,3-Dioxan-4-ol, 2,6-dimethyl-, acetate (9CI), m-Dioxan-4-ol, 2,6-dimethyl-, acetate (7CI,8CI), 2,4-Dimethyl-6-acetoxy-1,3-dioxane, 2,4-Dimethyl-6-m-dioxanyl acetate, 2,6-Dimethyl-m-dioxan-4-ol acetate, 2,6-Dimethyl-m-dioxan-4-yl acetate, 6-Acetoxy-2,4-dimethyl-1,3-dioxane, 6-Acetoxy-2,4-dimethyl-m-dioxane, 6-Acetoxy-2,4-dimethyl-m-dioxane, Acetomethoxane, Bioban DXN, Dimethoxane, Dioxin, Dioxin (bactericide), Dioxin CO, Giv Gard DXN, Giv Gard DXN-CO
IUPAC Name (2,6-dimethyl-1,3-dioxan-4-yl) acetate
SMILES CC1CC(OC(=O)C)OC(C)O1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H14O4/c1-5-4-8(11-6(2)9)12-7(3)10-5/h5,7-8H,4H2,1-3H3
Formula Weight 174.0892
Chemical Name or Material Dimethoxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.