missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethomorph 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Dimethomorph |
| CAS | 110488-70-5 |
| Analyte or Component Names | Dimethomorph |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H22 Cl N O4 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C21H22ClNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3/b18-14+ |
| SMILES | COc1ccc(cc1OC)\C(=C\C(=O)N2CCOCC2)\c3ccc(Cl)cc3 |
| IUPAC Name | (E)-3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-morpholin-4-ylprop-2-en-1-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction