missing translation for 'onlineSavingsMsg'
Learn More

Dimethoate, TRC

Product Code. 17947339
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947339 1 g 1g
17938089 500 mg 500mg
17968539 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17947339 Supplier Toronto Research Chemicals Supplier No. TRCD4605051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-51-5
Formula Weight 228.9996
InChI Formula InChI=1S/C5H12NO3PS2/c1-6-5(7)4-12-10(11,8-2)9-3/h4H2,1-3H3,(H,6,7)
IUPAC Name 2-dimethoxyphosphinothioylsulfanyl-N-methylacetamide
Molecular Formula C5 H12 N O3 P S2
Molecular Weight (g/mol) 229.26
SMILES CNC(=O)CSP(=S)(OC)OC
Synonym Phosphorodithioic acid, O,O-dimethyl S-(methylcarbamoylmethyl) ester (6CI), Phosphorodithioic acid, O,O-dimethyl ester, S-ester with 2-mercapto-N-methylacetamide (8CI), 8014 Bis HC, Aadimethoal, Agrithoate 40EC, American Cyanamid 12,880, BI 58, Bi 58 Nowy, Bi 58 Nowy 400EC, CL 12880, Cerox, Chloromezyl, Cygon, Cygon 2E, Cygon 400, Cygon 4E, Cygon Insecticide, Danadim, Daphene, De-fend, Dimate, Dimate (insecticide), Dimethoate, Dimeton, Dimevur, Dimezyl, Dimezyl 400EC, Ditimur 40, Dudu Ethoate, ENT 24650, Experimental Insecticide 12880, FIP, Fosfatox R, Fosfotox, Fosfotox R, Fosfotox R 35, Fostion MM, Laition, Lurgo, O,O-Dimethyl S-(N-methylcarbamoylmethyl) dithiophosphate, O,O-Dimethyl S-(N-methylcarbamoylmethyl) phosphorodithioate, PEI 75, Perfekthion, Perfekthion S, Phosphamid, Phosphamide, Racusan, Rogor, Rogor 20L, Rogor 40, Rogor L, Rogor P, Roxion, S-Methylcarbamoylmethyl O,O-dimethyl phosphorodithioate, Sinoratox, Sistemin, Super Ethoate, Systemin, Systemin (pesticide), Systoate, TATA TAFGOR, Tafgor 40EC, Tara, Tara 909, Tiomet
Chemical Name or Material Dimethoate

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.