missing translation for 'onlineSavingsMsg'
Learn More

Dimethoate, Fluka™

Product Code. 16292852 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16292852 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16292852 Supplier Honeywell-Fluka Supplier No. R1625100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Analytical standard, reference material

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethoate
Packaging Glass Bottle
CAS 60-51-5
Melting Point 46°C to 48°C
Boiling Point 117°C (0.1 hPa)
Synonym O,O-Dimethyl-S-methylcarbamoylmethyl-phosphordithioat Dithiophosphoric acid-O,O-dimethyl-S-(N-methylcarbamoylmethyl)-ester
Industry Type Environmental Analysis
Molecular Weight (g/mol) 229.26
Quantity 100 mg
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.