missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethindene Maleate, TRC
Specifications
Specifications
| Chemical Name or Material | Dimetindene maleate |
| CAS | 3614-69-5 |
| Molecular Formula | C20 H24 N2 . C4 H4 O4 |
| Quantity | 10 mg |
| Synonym | Dimetindene maleate, N,N-Dimethyl-2-[3-[(RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine (Z)-butenedioate, 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (2Z)-2-butenedioate (1:1), 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (Z)-2-butenedioate (1:1), Pyridine, 2-[1-[2-(dimethylaminoethyl)inden-3-yl]ethyl]-, maleate (6CI), Pyridine, 2-[1-[2-[2-(dimethylamino)ethyl]inden-3-yl]ethyl]-, maleate (1:1) (7CI,8CI), 2-[1-[2-(2-Dimethylaminoethyl)inden-3-yl]ethyl]pyridine maleate, Dimethinden maleate, Dimethindene maleate, Dimethpyrindene maleate, Fenistil, Fenistil-retard, Fenostil, Forhistal maleate, NSC 107677, Su-6518 |
| InChI Formula | InChI=1S/C20H24N2.C4H4O4/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2)3)14-16-8-4-5-9-18(16)20;5-3(6)1-2-4(7)8/h4-10,12,15H,11,13-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CC(C1=C(CCN(C)C)Cc2ccccc12)c3ccccn3.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N,N-dimethyl-2-[3-(1-pyridin-2-ylethyl)-1H-inden-2-yl]ethanamine |
| Molecular Weight (g/mol) | 408.49 |
| Formula Weight | 408.2049 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?