missing translation for 'onlineSavingsMsg'
Learn More

Dimethindene Maleate, TRC

Product Code. 30386704
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386704 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386704 Supplier Toronto Research Chemicals Supplier No. TRCD4604601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimetindene maleate
CAS 3614-69-5
Molecular Formula C20 H24 N2 . C4 H4 O4
Quantity 1 mg
Synonym Dimetindene maleate, N,N-Dimethyl-2-[3-[(RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine (Z)-butenedioate, 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (2Z)-2-butenedioate (1:1), 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (Z)-2-butenedioate (1:1), Pyridine, 2-[1-[2-(dimethylaminoethyl)inden-3-yl]ethyl]-, maleate (6CI), Pyridine, 2-[1-[2-[2-(dimethylamino)ethyl]inden-3-yl]ethyl]-, maleate (1:1) (7CI,8CI), 2-[1-[2-(2-Dimethylaminoethyl)inden-3-yl]ethyl]pyridine maleate, Dimethinden maleate, Dimethindene maleate, Dimethpyrindene maleate, Fenistil, Fenistil-retard, Fenostil, Forhistal maleate, NSC 107677, Su-6518
InChI Formula InChI=1S/C20H24N2.C4H4O4/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2)3)14-16-8-4-5-9-18(16)20;5-3(6)1-2-4(7)8/h4-10,12,15H,11,13-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES CC(C1=C(CCN(C)C)Cc2ccccc12)c3ccccn3.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;N,N-dimethyl-2-[3-(1-pyridin-2-ylethyl)-1H-inden-2-yl]ethanamine
Molecular Weight (g/mol) 408.49
Formula Weight 408.2049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.