missing translation for 'onlineSavingsMsg'
Learn More

Dimethenamid, TRC

Product Code. 17998879
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998879 100 mg 100mg
17948899 10 mg 10mg
17938159 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998879 Supplier Toronto Research Chemicals Supplier No. TRCD460580100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87674-68-8
Molecular Formula C12 H18 Cl N O2 S
Molecular Weight (g/mol) 275.79
Synonym 2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-(2-methoxy-1-methylethyl)acetamide, Dimethenamid, Fieldstar, Frontier, SAN 582, SAN 582H
IUPAC Name 2-chloro-N-(2,4-dimethylthiophen-3-yl)-N-(1-methoxypropan-2-yl)acetamide
SMILES COCC(C)N(C(=O)CCl)c1c(C)csc1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H18ClNO2S/c1-8-7-17-10(3)12(8)14(11(15)5-13)9(2)6-16-4/h7,9H,5-6H2,1-4H3
Formula Weight 275.0747
Chemical Name or Material Dimethenamid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.