missing translation for 'onlineSavingsMsg'
Learn More

Dimethenamid-d3, TRC

Product Code. 30384753
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384753 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30384753 Supplier Toronto Research Chemicals Supplier No. TRCD46058210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethenamid-d3
CAS 1246816-31-8
Purity Grade Notes HPLC
Molecular Formula C12H15D3ClNO2S
Quantity 10 mg
Synonym 2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-[2-(methoxy-d3)-1-methylethyl]acetamide, Fieldstar-d3, Frontier-d3, SAN 582-d3, SAN 582H-d3
InChI Formula InChI=1S/C12H18ClNO2S/c1-8-7-17-10(3)12(8)14(11(15)5-13)9(2)6-16-4/h7,9H,5-6H2,1-4H3/i4D3
SMILES O=C(CCl)N(C(C)COC([2H])([2H])[2H])C1=C(C)SC=C1C
Molecular Weight (g/mol) 278.81
Formula Weight 278.0935
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.