missing translation for 'onlineSavingsMsg'
Learn More

Dimethachlor Oxalic Acid Ethyl Ester, TRC

Product Code. 30387222
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387222 500 mg 500mg
30382919 100 mg 100mg
30382606 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387222 Supplier Toronto Research Chemicals Supplier No. TRCD467135500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethachlor Oxalic Acid Ethyl Ester
Analyte or Component Names Dimethachlor Oxalic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C15 H21 N O4
Synonym Ethyl 2-((2,6-Dimethylphenyl)(2-methoxyethyl)amino)-2-oxoacetate
InChI Formula InChI=1S/C15H21NO4/c1-5-20-15(18)14(17)16(9-10-19-4)13-11(2)7-6-8-12(13)3/h6-8H,5,9-10H2,1-4H3
Solution Type Neat
SMILES O=C(OCC)C(N(C1=C(C)C=CC=C1C)CCOC)=O
IUPAC Name ethyl 2-[N-(2-methoxyethyl)-2,6-dimethylanilino]-2-oxoacetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 279.332
Quantity 500 mg
Formula Weight 279.147
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.