missing translation for 'onlineSavingsMsg'
Learn More

Dimesna, TRC

Product Code. 30373017
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373017 100 mg -
30374003 250 mg 250mg
30387419 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373017 Supplier Toronto Research Chemicals Supplier No. TRCD460660100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16208-51-8
Molecular Formula C4 H8 O6 S4 . 2 Na
Molecular Weight (g/mol) 326.34
Synonym 2,2'-Dithiobis-ethanesulfonic Acid Sodium Salt (1:2), Ethanesulfonic acid, 2,2'-dithiobis-, disodium salt (9CI), Ethanesulfonic acid, 2,2'-dithiodi-, disodium salt (6CI,8CI), 2,2'-Dithiodi-1-ethanesulfonic acid disodium salt, BNP 7787, Bis(2-sulfoethyl)disulfide disodium salt, Dimesna, Disodium 2,2'-dithiobis(ethanesulfonate), Disodium 2,2'-dithiodiethanesulfonate, NSC 716976, Tavocept
IUPAC Name disodium;2-(2-sulfonatoethyldisulfanyl)ethanesulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)CCSSCCS(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H10O6S4.2Na/c5-13(6,7)3-1-11-12-2-4-14(8,9)10;;/h1-4H2,(H,5,6,7)(H,8,9,10);;/q;2*+1/p-2
Formula Weight 325.8999
Chemical Name or Material Dimesna
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.