missing translation for 'onlineSavingsMsg'
Learn More

Dimepranol-d6, TRC

Product Code. 30388002
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388002 5 mg
30370088 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388002 Supplier Toronto Research Chemicals Supplier No. TRCD4604025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97964-89-1
Molecular Formula C5H7D6NO
Molecular Weight (g/mol) 109.2
Synonym (±)-1-[(Dimethyl-d6)amino]-2-propanol, (±)-1-(N,N-Dimethyl-d6)amino]-2-propanol, 1-[(Dimethyl-d6)amino]-2-propanol, 1-Methyl-2-[(dimethyl-d6)amino]ethanol, 2-[(Dimethyl-d6)amino]-1-methyl-1-ethanol, 2-[(Dimethyl-d6)amino]-1-methylethanol, 3-[(Dimethyl-d6)amino]-2-propanol, Bisomer Amine D 700, DL-1-[(Dimethyl-d6)amino]-2-propanol, DMA 2P, (Dimethyl-d6)(2-hydroxypropyl)amine, (Dimethyl-d6)isopropanolamine, N,N-(Dimethyl-d6)-2-hydroxy-1-propanamine, N,N-(Dimethyl-d6)-2-hydroxypropylamine, N,N-(Dimethyl-d6)amino-2-propanol, N,N-(Dimethyl-d6)isopropanolamine, NSC 3163
SMILES OC(CN(C([2H])([2H])[2H])C([2H])([2H])[2H])C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3/i2D3,3D3
Formula Weight 109.1374
Chemical Name or Material Dimepranol-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.