missing translation for 'onlineSavingsMsg'
Learn More

Dimedone, TRC

Product Code. 30382838
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382838 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382838 Supplier Toronto Research Chemicals Supplier No. TRCD46020025G
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimedone
CAS 126-81-8
Molecular Formula C8H12O2
Quantity 25 g
Synonym 5,5-Dimethyl-1,3-cyclohexanedione, 1,1-Dimethyl-3,5-cyclohexanedione, 1,1-Dimethyl-3,5-diketocyclohexane, 5,5-Dimethyl-1,3-cyclohexandione, 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethyldihydroresorcinol, 5,5-Dimethylhydroresorcinol, Cyclomethone, Dimedon, Lu 274, Medon, Methon, Methone, NSC 14984, NSC 17544, NSC 242994, NSC 43759
InChI Formula InChI=1S/C8H12O2/c1-8(2)4-6(9)3-7(10)5-8/h3-5H2,1-2H3
SMILES CC1(C)CC(=O)CC(=O)C1
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
Molecular Weight (g/mol) 140.18
Formula Weight 140.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.