missing translation for 'onlineSavingsMsg'
Learn More

Dimedone, 98%

Product Code. 11479402
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479402 100 g 100g
11469402 25 g 25g
11489402 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479402 Supplier Thermo Scientific Alfa Aesar Supplier No. A10140.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126-81-8 | C8H12O2 | 140.18 g/mol

Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used in synthesis of fused 3-methylfuransDimedone is used in organic chemistry to identify the presence of aldehyde group. It is also used as a catalyst in the formation of transition-metal complexes. Further, it is also employed in colorimetry, crystallography, luminescence and spectrophotometric analysis. In addition to this, it is used in the synthesis of naturally occurring fused 3-methylfurans.

Solubility
Soluble in methanol, ethanol, benzene and chloroform. Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-81-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001588
InChI Key BADXJIPKFRBFOT-UHFFFAOYSA-N
Synonym dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
PubChem CID 31358
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(C)CC(=O)CC(=O)C1

Specifications

Melting Point 146°C to 150°C
Quantity 100 g
Beilstein 471489
Merck Index 14,3242
Solubility Information Soluble in methanol,ethanol,benzene and chloroform. Slightly soluble in water.
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material Dimedone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.