missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Various Instruments

Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for Various Instruments has the knowledge and expertise that goes into manufacturing an SGE syringe.

Brand:  Trajan Scientific and Medical™ 008987

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258513

  • £109.00 / Each
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for various instruments
Specifications

Specifications

60mm stroke, 6-32 UNC thread in plunger button
60 mm
10 mL
Various Instruments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers