Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Product Code. 15218483
Change view
Click to view available options
Volume (Metric):
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15218483 50 μL Each
15238473 25 μL Each
15238493 250 μL Each
15238513 5 mL Each
15248503 1 mL Each
15278513 10 mL Each
15288483 100 μL Each
15288503 2.5 mL Each
15288513 25 mL Each
15298493 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15218483 Supplier Trajan Scientific and Medical™ Supplier No. 004696
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V6 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for V6 Pumps
Specifications
Description 60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger
Volume (Metric) 50 μL
For Use With (Equipment) V6 Pump