Learn More
Thermo Scientific™ DiluFlux Integral Stand for Broth Bags
Product Code. 17173784
Change view
Click to view available options
For Use With (Equipment):
DiluFlux Sample Dilution
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17173784 DiluFlux Sample Dilution Each
1 options
Product Code. 17173784 Supplier Thermo Scientific™ Supplier No. DB4109A
  • ONLINE EXCLUSIVE PRICE
    £404.00
    £370.55 / Pack of 1
    Save £33.45
    8% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Lägg i varukorgen
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
DiluFlux Stand for bagged media. Integrated to the unit, to hold one broth bag. Delivered with 4 screws + Torx screwdriver
Compatible with many prepared media formats, theeasy-to-clean, stainless steel DiluFlux Pro Dilutors are an ideal fit for any food testing laboratory.
Automated media dispensing and sample dilution
  • Compact and easy-to-use
  • Automated, rapid and accurate dispensing
  • Available with one or two pump heads for multiple media dispensing
  • Compatible with Dry-Bags and other media formats
TRUSTED_SUSTAINABILITY
Specifikationer
Product Type Bag Stand
For Use With (Equipment) DiluFlux Sample Dilution
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.