Learn More
Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Click to view available options
Product Type:
Nozzles
Trocar
Packaging Quantity:
1 Ea.
5 pieces/Bag
Unit Size:
Each
Pack of 5
Description
Achieve maximum productivity with the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories. Expand the flexibility and maintain long-term functionality of the two platforms with accessories that enable the use of different media formats (bottles/bags), enhance the user interface and support proper function of the system.
The easy-to-use Diluflux Gravimetric Dilutor allows the simultaneous use of up to two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted on the unit. The reliable and powerful Homogenizer Laboratory Blender enables rapid and efficient sample homogenization with a small footprint and quiet operation. The accessories listed here enable:
Not all products are available for sale in all territories. Please inquire.
- Alternative receptacles (bottles instead of bags)
- Alternative media sources (broth bags/bottles)
- Additional/replacement bag stands for ease of preparation
- Bag storage for incubation
- Sample labeling
- Maintenance with replacement parts
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Product Type | Trocar |
| For Use With (Equipment) | Diluflux™ Diluter and Homogenizer |
| Packaging Quantity | 1 Ea. |
For research use only.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido