Learn More
Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Product Code. 17153784
Change view
Click to view available options
Product Type:
Nozzles
Trocar
Packaging Quantity:
1 Ea.
5 pieces/Bag
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Packaging Quantity unitSize
17153784 Trocar 1 Ea. Each
17163784 Nozzles 5 pieces/Bag Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17153784 Supplier Thermo Scientific™ Supplier No. DB4002A
  • ONLINE EXCLUSIVE PRICE
    £191.00
    £186.75 / Pack of 1
    Save £4.25
    2% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Accurately dispense media and achieve powerful, quiet, reliable homogenization with maximum productivity in your sample preparation.
Achieve maximum productivity with the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories. Expand the flexibility and maintain long-term functionality of the two platforms with accessories that enable the use of different media formats (bottles/bags), enhance the user interface and support proper function of the system.
The easy-to-use Diluflux Gravimetric Dilutor allows the simultaneous use of up to two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted on the unit. The reliable and powerful Homogenizer Laboratory Blender enables rapid and efficient sample homogenization with a small footprint and quiet operation. The accessories listed here enable:
  • Alternative receptacles (bottles instead of bags)
  • Alternative media sources (broth bags/bottles)
  • Additional/replacement bag stands for ease of preparation
  • Bag storage for incubation
  • Sample labeling
  • Maintenance with replacement parts
Contact us to arrange a demonstration of the Dilutor, the Blender, and/or its accessories.
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Trocar
For Use With (Equipment) Diluflux™ Diluter and Homogenizer
Packaging Quantity 1 Ea.