missing translation for 'onlineSavingsMsg'
Learn More

Diltiazem hydrochloride, MedChemExpress

Product Code. 16458727
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458727 5 g 5g
16401439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458727 Supplier Medchem Express Supplier No. HY14656
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diltiazem hydrochloride is a Ca2+ influx inhibitor (slow channel blocker or calcium antagonist).

Diltiazem hydrochloride is a Ca2+ influx inhibitor (slow channel blocker or calcium antagonist).

Specifications

Chemical Name or Material Diltiazem hydrochloride
CAS 33286-22-5
Color White
Purity Grade Notes Research
Molecular Formula C22H27ClN2O4S
Quantity 5 g
Synonym CRD-401
Solubility Information H2O : 33.33 mg/mL (73.91 mM; Need ultrasonic)
SMILES CN(C)CCN1C2=CC=CC=C2S[C@@H](C3=CC=C(OC)C=C3)[C@@H](OC(C)=O)C1=O.Cl
Molecular Weight (g/mol) 450.98
Formula Weight 450.98
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.