missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein/Oregon Green Biotin Polyclonal Antibody, Invitrogen™

Fluorescein/Oregon Green Antibody, Biotin conjugate, A-982, from Invitrogen™. Species Reactivity: Chemical; Applications: Flow Cytometry, Immunocytochemistry, Immunohistochemistry.

Brand:  Invitrogen A982

Product Code. 10276162

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Fluorescein/Oregon Green Antibody, Biotin conjugate, A-982, from Invitrogen™. Species Reactivity: Chemical; Applications: Flow Cytometry, Immunocytochemistry, Immunohistochemistry.

Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
Host Species: Rabbit
Cross Reactivity: Chemical
Isotype: IgG
Immunogen: Fluorescein/Oregon Green
Formulation: 500μL purified antibody (1mg/mL) in 0.1M potassium phosphate and 5mM sodium azide; pH 8
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescein/Oregon Green
Polyclonal
500μL Purified Antibody (1mg/ml) in 0.1M potassium phosphate, with 5mM sodium azide; pH8
Fluorescein/Oregon Green
Primary
Purified
Chemical
Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Biotin
Rabbit
500 μL
Store at 4° C
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.