missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dilithium Tetrachlorocuprate (0.1M solution in THF), TRC
Specifications
Specifications
| Chemical Name or Material | Dilithium Tetrachlorocuprate |
| CAS | 15489-27-7 |
| Molecular Formula | Cl4CuLi22- |
| Quantity | 250 mg |
| Synonym | Dilithium Tetrachlorocuprate(2-), Dilithium Copper Tetrachloride, Copper Lithium Chloride |
| InChI Formula | InChI=1S/4ClH.Cu.2Li/h4*1H;;;/q;;;;+2;2*+1/p-4 |
| SMILES | [Li+].[Li+].[Cl-][Cu+2]([Cl-])([Cl-])[Cl-] |
| IUPAC Name | Lithium Tetrachlorocuprate(II) |
| Molecular Weight (g/mol) | 219.24 |
| Formula Weight | 216.84 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?