missing translation for 'onlineSavingsMsg'
Learn More

Dilithium Tetrachlorocuprate (0.1M solution in THF), TRC

Product Code. 30372975
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372975 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372975 Supplier Toronto Research Chemicals Supplier No. TRCD459905100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dilithium Tetrachlorocuprate
CAS 15489-27-7
Molecular Formula Cl4CuLi22-
Quantity 100 mg
Synonym Dilithium Tetrachlorocuprate(2-), Dilithium Copper Tetrachloride, Copper Lithium Chloride
InChI Formula InChI=1S/4ClH.Cu.2Li/h4*1H;;;/q;;;;+2;2*+1/p-4
SMILES [Li+].[Li+].[Cl-][Cu+2]([Cl-])([Cl-])[Cl-]
IUPAC Name Lithium Tetrachlorocuprate(II)
Molecular Weight (g/mol) 219.24
Formula Weight 216.84
Concentration or Composition (by Analyte or Components) 0.1 M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.