missing translation for 'onlineSavingsMsg'
Learn More

Dilithium tetrachlorocuprate, 0.1M solution in THF, AcroSeal™

Product Code. 10587380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10587380 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10587380 Supplier Thermo Scientific Acros Supplier No. 429051000
  • ONLINE EXCLUSIVE PRICE
    £63.45
    £55.65 / 100mL
    Save £7.80
    12% Off
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15489-27-7 | Cl4CuLi2 | 219.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dilithium tetrachlorocuprate
CAS 15489-27-7, 109-99-9
Quantity 100 mL
Molecular Formula Cl4CuLi2
Linear Formula Li2CuCl4
MDL Number MFCD00011081
Flash Point −17°C
InChI Key HCJWWBBBSCXJMS-UHFFFAOYSA-J
SMILES [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
IUPAC Name dilithium;tetrachlorocopper(2-)
Molecular Weight (g/mol) 219.24
PubChem CID 11074879
Formula Weight 219.24
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.91
Density 0.9100g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dilithium tetrachlorocuprate, 0.1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.