missing translation for 'onlineSavingsMsg'
Learn More

Dilauroyl peroxide, 97%

Product Code. 11303466
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303466 500 g 500g
11393456 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303466 Supplier Thermo Scientific Alfa Aesar Supplier No. L14310.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-74-8 | C24H46O4 | 398.628 g/mol

Lauroyl peroxide acts as a bleaching agent, drying agent for fats, oils and waxes. Further, it serves as a polymerization initiator as well as vulcanizing agent. In addition to this, it plays an important role for high-pressure polyethylene and food used in bleaching agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lauroyl peroxide acts as a bleaching agent, drying agent for fats, oils and waxes. Further, it serves as a polymerization initiator as well as vulcanizing agent. In addition to this, it plays an important role for high-pressure polyethylene and food used in bleaching agent.

Solubility
Soluble in oils and organic solvents. Slightly soluble in alcohol. Insoluble in water.

Notes
Incompatible with acids, bases, metals, reducing agents, powdered metals, strong reducing agents and combustible material.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-74-8
Molecular Formula C24H46O4
Molecular Weight (g/mol) 398.628
MDL Number MFCD00008964
InChI Key YIVJZNGAASQVEM-UHFFFAOYSA-N
Synonym lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle
PubChem CID 7773
ChEBI CHEBI:82479
IUPAC Name dodecanoyl dodecaneperoxoate
SMILES CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC

Specifications

Melting Point 53°C to 57°C
Density 0.91
Quantity 500 g
UN Number UN3106
Beilstein 1804936
Solubility Information Soluble in oils and organic solvents. Slightly soluble in alcohol. Insoluble in water.
Formula Weight 398.63
Percent Purity 97%
Chemical Name or Material Lauroyl peroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.