missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylphosphine, 10 wt.% solution in hexanes

Product Code. 10756183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10756183 10 g Glass bottle 10g
10539613 2.5 g Glass bottle 2.5g
10005752 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10756183 Supplier Thermo Scientific Acros Supplier No. 383400100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20491-53-6 | C6H15P | 118.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropylphosphine
CAS 20491-53-6, 110-54-3
Color Brown-Yellow to Yellow
Density 0.6700g/mL
Flash Point −22°C
Packaging Glass bottle
Molecular Formula C6H15P
Linear Formula [(CH3)2CH]2PH
MDL Number MFCD00233814
Quantity 10 g
Specific Gravity 0.67
Synonym diisopropylphosphine, di-isopropylphosphine, di-i-propylphosphine, diisopropylphosphane, phosphine, diisopropyl, phosphine, bis 1-methylethyl, ipr2ph, di-iso-propylphosphine, di-iso-propyl phosphine, di propan-2-yl phosphane
Solubility Information Solubility in water: insoluble
InChI Key WDIIYWASEVHBBT-UHFFFAOYSA-N
SMILES CC(C)PC(C)C
IUPAC Name di(propan-2-yl)phosphane
Molecular Weight (g/mol) 118.16
PubChem CID 536496
Formula Weight 118.16
Percent Purity 9 to 11% (Total base)
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisopropylphosphine, 10 wt.% solution in hexanes
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.