missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine Hydrochloride, TRC

Product Code. 30386146
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386146 250 mg 250mg
30378281 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386146 Supplier Toronto Research Chemicals Supplier No. TRCD493140250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropylamine Hydrochloride
CAS 819-79-4
Molecular Formula C6H15N • HCl
Quantity 250 mg
Synonym 2-Propanamine, N-(1-methylethyl)-, hydrochloride (1:1) (ACI), 2-Propanamine, N-(1-methylethyl)-, hydrochloride (9CI), Diethylamine, dimethyl-, hydrochloride (6CI), Diisopropylamine, hydrochloride (8CI), Bis(isopropyl)ammonium chloride, Diisopropylammonium chloride, N-(1-Methylethyl)-2-propanamine hydrochloride, Diisopropylamine hydrochloride, N,N-Diisopropylamine hydrochloride
InChI Formula InChI=1S/C6H15N.ClH/c1-5(2)7-6(3)4;/h5-7H,1-4H3;1H
SMILES CC(C)NC(C)C.Cl
Molecular Weight (g/mol) 137.65
Formula Weight 137.0971
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.