missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl sulfide, 99%

Product Code. 11499207
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499207 10 g 10g
11409217 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499207 Supplier Thermo Scientific Alfa Aesar Supplier No. B21302.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 625-80-9 | C6H14S | 118.24 g/mol

Used as a flavoring agents and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agents and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-80-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00008895
InChI Key XYWDPYKBIRQXQS-UHFFFAOYSA-N
Synonym diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
PubChem CID 12264
SMILES CC(C)SC(C)C

Specifications

Density 0.814
Boiling Point 119°C to 120°C
Flash Point 7°C (44°F)
Odor Stench
Linear Formula (CH3)2CHSCH(CH3)2
Refractive Index 1.4385
Quantity 10 g
UN Number UN1993
Beilstein 1731257
Solubility Information Not miscible or difficult to mix with water.
IUPAC Name 2-propan-2-ylsulfanylpropane
Formula Weight 118.24
Percent Purity 99%
Chemical Name or Material Diisopropyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.